missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) oxalate dihydrate, Mn 30% min

As a paint and varnish drier

Manufacturer:  Alfa Aesar™ 1468506

 View more versions of this product

Catalog No. AA1468506



Specifications

Specifications

Manganese(II) oxalate dihydrate
Odorless
100°C (decomposition)
6556-16-7
MFCD00150452
manganese ii oxalate 2-hydrate
Soluble in dilute acids; Slightly soluble in water
C(=O)(C(=O)[O-])[O-].O.O.[Mn+2]
178.986
178.99 (142.96 Anhydrous)
2.453g/mL
White
5g
C2H4MnO6
14,5734
Mn 30% min.
HDJUVFZHZGPHCQ-UHFFFAOYSA-L
manganese(2+);oxalate;dihydrate
131882857
Powder/Chunks

Chemical Identifiers

6556-16-7
178.986
HDJUVFZHZGPHCQ-UHFFFAOYSA-L
131882857
C(=O)(C(=O)[O-])[O-].O.O.[Mn+2]
C2H4MnO6
MFCD00150452
manganese ii oxalate 2-hydrate
manganese(2+);oxalate;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents