missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) nitrate hydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1080618

 View more versions of this product

Catalog No. AA1080618



Specifications

Specifications

Manganese(II) nitrate hydrate
99.999% (Metal basis)
MFCD00149788
acmc-1brrd
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Mn+2]
196.961
178.95 (anhydrous)
Odorless
50g
15710-66-4
H2MnN2O7
Pink Crystalline Aggregates
HBTFASPVVFSRRI-UHFFFAOYSA-N
manganese(2+);dinitrate;hydrate
16211480
Crystalline Aggregates
Pink

Chemical Identifiers

15710-66-4
196.961
HBTFASPVVFSRRI-UHFFFAOYSA-N
16211480
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Mn+2]
H2MnN2O7
MFCD00149788
acmc-1brrd
manganese(2+);dinitrate;hydrate
Safety and Handling

Safety and Handling

H272-H301-H332-H315-H319-H335; May intensify fire; oxidizer. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: III

Documents