missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) nitrate hydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1080609

 View more versions of this product

Catalog No. AA1080609



Specifications

Specifications

Manganese(II) nitrate hydrate
99.999% (Metal basis)
MFCD00149788
acmc-1brrd, manganese 2+ hydrate dinitrate, manganese 2+ ion hydrate dinitrate, manganese 2+ nitrate-water 1/2/1, manganese ii nitrate hydrate, manganese ii nitrate hydrate, puratronic, manganese nitrate hydrate, mn.2no3.h2o
HBTFASPVVFSRRI-UHFFFAOYSA-N
manganese(2+);dinitrate;hydrate
16211480
Crystalline Aggregates
Hygroscopic
∽26°C
15710-66-4
H2MnN2O7
Pink Crystalline Aggregates
Soluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Mn+2]
196.961
178.95 (Anhydrous)
Odorless
Pink
10g

Chemical Identifiers

15710-66-4
196.961
HBTFASPVVFSRRI-UHFFFAOYSA-N
16211480
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Mn+2]
H2MnN2O7
MFCD00149788
acmc-1brrd, manganese 2+ hydrate dinitrate, manganese 2+ ion hydrate dinitrate, manganese 2+ nitrate-water 1/2/1, manganese ii nitrate hydrate, manganese ii nitrate hydrate, puratronic, manganese nitrate hydrate, mn.2no3.h2o
manganese(2+);dinitrate;hydrate
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H272-H302-H314-H373

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-828-8

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Documents