Please login to your online account to display your discounted pricing

Manganese(II) nitrate hydrate, 99.98% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Manganese(II) nitrate hydrate
CAS 15710-66-4
Physical Form Lump
Assay Percent Range 99.98% (metals basis)
Molecular Formula Mn(NO3)2·xH2O
Formula Weight 178.95
MDL Number MFCD00149788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8784822

  SDS
alfa aesar
8784822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8784836

  SDS
alfa aesar
8784836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.95
Physical Form Lump
Quantity 100g
Color Pink
DOT Information DOT Class: 5.1, Packaging Group: III
EINECS Number 233-828-8
CAS 15710-66-4
Health Hazard 1 H272-H301-H332-H315-H319-H335: May intensify fire; oxidizer. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Odor Odorless
Molecular Formula Mn(NO3)2·xH2O
Assay Percent Range 99.98% (metals basis)
Chemical Name or Material Manganese(II) nitrate hydrate
MDL Number MFCD00149788
Safety and Handling

H272-H301-H332-H315-H319-H335: May intensify fire; oxidizer. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.