missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) methoxide

Manufacturer:  Alfa Aesar™ 2256306

 View more versions of this product

Catalog No. AA2256306



Specifications

Specifications

Manganese(II) methoxide
Undesignated
7245-20-7
Mn(OCH3)2
UN3181
Soluble in methanol
C[O-].C[O-].[Mn+2]
117.006
117.01
Moisture Sensitive
5g
C2H6MnO2
MFCD00061476
manganese ii methoxide, acmc-20akkm, dimethoxymanganese ii, manganese ii bis methoxide, manganese 2+ bis methoxide, manganese 2+ ion bis methoxide
UQIRCVPINUNHQY-UHFFFAOYSA-N
manganese(2+);methanolate
10219413
Powder

Chemical Identifiers

7245-20-7
117.006
UQIRCVPINUNHQY-UHFFFAOYSA-N
10219413
C[O-].C[O-].[Mn+2]
C2H6MnO2
MFCD00061476
manganese ii methoxide, acmc-20akkm, dimethoxymanganese ii, manganese ii bis methoxide, manganese 2+ bis methoxide, manganese 2+ ion bis methoxide
manganese(2+);methanolate
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280h-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

SDS Documents