missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) iodide, ultra dry, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4716606

 View more versions of this product

Catalog No. AA4716606



Description

Description

  • Ampouled under argon
Specifications

Specifications

Manganese(II) iodide
7790-33-2
MFCD00016225
308.75
99.99%
−10 Mesh
Ampule
Red to White
5g
Ultra dry
(metals basis)
14,5732
Beads
5.0g/mL
Odorless
Light, Air & Moisture Sensitive
638°C

Chemical Identifiers

7790-33-2
MFCD00016225
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents