Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) iodide tetrahydrate, 98%

$84.82 - $317.93

Chemical Identifiers

Chemical Name or Material Manganese(II) iodide tetrahydrate
CAS 13446-37-2
Molecular Formula MnI2·4H2O
Formula Weight 380.81 (308.75 Anhydrous)
MDL Number MFCD00150257
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1764909

  SDS
alfa aesar™
A1764909
Each for $84.82

AAA1764918

  SDS
alfa aesar™
A1764918
Each for $317.93
Description & Specifications

Specifications

Formula Weight 380.81 (308.75 Anhydrous)
Quantity 10g
CAS 13446-37-2
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Molecular Formula MnI2·4H2O
MDL Number MFCD00150257
TSCA TSCA
Health Hazard 1 Warning
Chemical Name or Material Manganese(II) iodide tetrahydrate
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.