Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) hexafluoro-2,4-pentanedionate trihydrate, 97%

Chemical Identifiers

Chemical Name or Material Manganese(II) hexafluoro-2,4-pentanedionate trihydrate
CAS 123334-26-9
Formula Weight 523.09g/mol (469.04g/mol Anhydrous)
MDL Number MFCD00150654
Synonym Manganese(II) hexafluoroacetylacetonate trihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5509403

 
alfa aesar™
H55094-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Yellow
Melting Point 173°C (dec.)
Quantity 1g
Chemical Name or Material Manganese(II) hexafluoro-2,4-pentanedionate trihydrate
CAS 123334-26-9
Physical Form Powder
Assay 97%
Formula Weight 523.09g/mol (469.04g/mol Anhydrous)
MDL Number MFCD00150654
Synonym Manganese(II) hexafluoroacetylacetonate trihydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.