Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) formate dihydrate, 98%

$192.86 - $192.86

Chemical Identifiers

Chemical Name or Material Manganese(II) formate dihydrate
CAS 3251-96-5
Molecular Formula Mn(HCO2)2·2H2O
Formula Weight 181.01 (144.98 Anhydrous)
MDL Number MFCD00050832
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1962222

  SDS
alfa aesar™
A1962222
100g Each for $192.86
Description & Specifications

Specifications

Formula Weight 181.01 (144.98 Anhydrous)
Chemical Name or Material Manganese(II) formate dihydrate
Assay 98%
Molecular Formula Mn(HCO2)2·2H2O
MDL Number MFCD00050832
CAS 3251-96-5
Solubility Information Soluble in water
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100g

Applications
Manganese(II) formate dihydrate is used for making dryer of ink and paint, catalyst of synthesis fatty acid, manganese compound, electrolyzed metallic manganese, dyeing and for printing and dyeing industries.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.