Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) chloride, ultra dry, 99.998% (metals basis)

Chemical Identifiers

Chemical Name or Material Manganese(II) chloride
CAS 7773-01-5
Name Note Ultra dry
Linear Formula MnCl2
Formula Weight 125.84
MDL Number MFCD00011114
UN Number UN3288
Merck Index 14,5728
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1469706

  SDS
alfa aesar™
1469706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1469714

  SDS
alfa aesar™
1469714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 125.84
Packaging Ampule
Chemical Name or Material Manganese(II) chloride
TSCA TSCA
Mesh Size −10 Mesh
Odor Odorless
Name Note Ultra dry
Physical Form Beads
Assay 99.998%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Linear Formula MnCl2
MDL Number MFCD00011114
CAS 7773-01-5
UN Number UN3288
Merck Index 14,5728
Boiling Point 1190°C
Color Pink
Melting Point 650°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 2.977g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
RTECS Number OO9625000

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.

  • Ampouled under argon