Please login to your online account to display your discounted pricing

Manganese(II) chloride tetrahydrofuran complex (1:2) 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC390460050

 View more versions of this product

Catalog No. AC390460050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 270.06
Quantity 5g
Color Light pink
Infrared Spectrum Authentic
Molecular Formula C8H16Cl2MnO2
Elemental Analysis 96.0% min.
Chemical Name or Material Manganese(II) chloride tetrahydrofuran complex (1:2)
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD12912454
CAS 70317-52-1
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.
Health Hazard 3 GHS P Statement:
Eliminate all ignition sources if safe to do so.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Manganese(II) chloride tetrahydrofuran complex (1:2)
Assay Percent Range 97%
Molecular Formula C8H16Cl2MnO2
Formula Weight 270.06
MDL Number MFCD12912454
Accessories

GHS Signal Word: Warning

GHS P Statement:
Eliminate all ignition sources if safe to do so.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.

SDS & Safety Documents