Please login to your online account to display your discounted pricing

Manganese(II) chloride tetrahydrofuran complex (1:2) 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Manganese(II) chloride tetrahydrofuran complex (1:2)
Assay Percent Range 97%
Molecular Formula C8H16Cl2MnO2
Formula Weight 270.06
MDL Number MFCD12912454
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390460010

  SDS
thermo fisher scientific
AC390460010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390460050

  SDS
thermo fisher scientific
AC390460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 270.06
Quantity 1g
Color Light pink
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Eliminate all ignition sources if safe to do so.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.
MDL Number MFCD12912454
Infrared Spectrum Authentic
Packaging Glass bottle
Elemental Analysis 96.0% min.
Molecular Formula C8H16Cl2MnO2
Assay Percent Range 97%
Chemical Name or Material Manganese(II) chloride tetrahydrofuran complex (1:2)
CAS 70317-52-1
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Eliminate all ignition sources if safe to do so.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.