Learn More
Manganese(II) chloride tetrahydrate, 99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01156336
Description
Manganese(II) chloride tetrahydrate is used in the tailing of DNA - MnCl2Â buffer is recommended for dC and dG tailing and in reverse transcription of RNA with thermostable DNA polymerase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Manganese(II) chloride tetrahydrate | |
13446-34-9 | |
Crystalline | |
99% (Metal basis) | |
MFCD00149792 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dichloride | |
197.90 (125.84 Anhydrous) | |
Hygroscopic | |
2.01 |
58°C | |
Pink | |
500 g | |
Cl2H8MnO4 | |
Soluble in pyridine,ethanol. Insoluble in ether. | |
O.O.O.O.[Cl-].[Cl-].[Mn++] | |
197.90 | |
Odorless | |
Pink Crystalline/powder |
Chemical Identifiers
13446-34-9 | |
197.90 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
O.O.O.O.[Cl-].[Cl-].[Mn++] |
Cl2H8MnO4 | |
MFCD00149792 | |
manganese(2+) tetrahydrate dichloride |
Safety and Handling
P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
H302-H318-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 231-869-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only