missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) chloride tetrahydrate, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 193451000

 View more versions of this product

Catalog No. AC193451000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Manganese(II) chloride tetrahydrate
13446-34-9
Cl2H8MnO4
MFCD00149792
15,5793
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese(2+) tetrahydrate dichloride
197.91
Glass bottle
58.0°C
100 g
MnCl2·4H2O
07,222; 09,288
Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
O.O.O.O.[Cl-].[Cl-].[Mn++]
197.90
ACS Reagent

Chemical Identifiers

13446-34-9
197.90
CNFDGXZLMLFIJV-UHFFFAOYSA-L
O.O.O.O.[Cl-].[Cl-].[Mn++]
Cl2H8MnO4
MFCD00149792
manganese(2+) tetrahydrate dichloride
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only