missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Manganese(II) Chloride Tetrahydrate, ≥99%, Reagent Grade, Honeywell™

Reagent Grade,≥99%

$96.78 - $96.78

Chemical Identifiers

CAS 13446-34-9
MDL Number MFCD00149792
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60048079 View Documents Honeywell™
M3634100G
100g Poly bottle Each for $96.78
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

13446-34-9
MFCD00149792

Specifications

13446-34-9
MFCD00149792
197.91g/mol
1.913g/cu.cm
Poly bottle
Crystalline
58°C
100g
MnCl2 · 4H2O
NONH for all modes of transport
≥99%
Odorless
Completely soluble
Pink
4.0 to 6.0 (at 20°C)
Manganese(II) chloride tetrahydrate
Safety and Handling

Safety and Handling

RTECSNumber : 231-869-6