missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) chloride tetrahydrate, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4444214

 View more versions of this product

Catalog No. AA4444214



Specifications

Specifications

Manganese (II) chloride tetrahydrate
(metals basis)
MFCD00149792
Soluble in water,acetic acid,pyridine,and ethanol but insoluble in ether.
Crystalline
2.010 g/mL
Hygroscopic
25g
13446-34-9
MnCl2·4H2O
14,5728
197.90 (125.84 Anhydrous)
99.99%
Odorless
58°C

Chemical Identifiers

13446-34-9
MnCl2·4H2O
MFCD00149792
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c

H302-H318-H373

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-869-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents