missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Manganese(Ii) Chloride Tetrahydrate, 98.0 to 101.0%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent,≥98%

$105.21 - $293.41

Chemical Identifiers

CAS 13446-34-9
MDL Number MFCD00149792
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045291 View Documents Honeywell Fluka™
221279100G
100g Poly Bottle Each for $105.21
 
60045292 View Documents Honeywell Fluka™
221279500G
500g Poly Bottle Each for $293.41
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

13446-34-9
MFCD00149792

Specifications

13446-34-9
MFCD00149792
197.91g/mol
ACS Reagent
100g
MnCl2 · 4H2O
NONH for all modes of transport
98.0 to 101.0%
Poly Bottle
Manganese(Ii) Chloride Tetrahydrate
Safety and Handling

Safety and Handling

P273

H302-H412

EINECSNumber : 231-869-6

RTECSNumber : OO9650000