missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 271410250

 View more versions of this product

Catalog No. AC271410250



Specifications

Specifications

Manganese(II) chloride
96% min. (Complexometry)
MnCl2
07,222
acmc-20akkt, ksc171o6h, manganese 2+ ion dichloride, manganese chloride, manganese ii chloride, manganese ii chloride anhydrous crystalline, manganesechloride, manganous chloride, anhydr, manganous chloride,anhydrous
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese(2+);dichloride
10313134
125.84
97%
90ppm max.
Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
1190.0°C
650.0°C
7773-01-5
Cl2Mn
MFCD00011114
15, 5793
Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol,insoluble in ether
[Cl-].[Cl-].[Mn+2]
125.84
CHEBI:63041
Flakes
200ppm max.
Glass bottle
1% max.
Pink
25g

Chemical Identifiers

7773-01-5
125.84
CNFDGXZLMLFIJV-UHFFFAOYSA-L
10313134
manganese(2+);dichloride
Cl2Mn
MFCD00011114
acmc-20akkt, ksc171o6h, manganese 2+ ion dichloride, manganese chloride, manganese ii chloride, manganese ii chloride anhydrous crystalline, manganesechloride, manganous chloride, anhydr, manganous chloride,anhydrous
CHEBI:63041
[Cl-].[Cl-].[Mn+2]
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using th

GHS Signal Word: Danger

EINECSNumber : 231-869-6

SDS Documents