missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) carbonate hydrate, 90%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 222540010

Catalog No. AC222540010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Manganese(II) carbonate hydrate
34156-69-9
1 kg
CMnO3
MFCD00149794
Solubility in water: insoluble. Other solubilities: soluble in dilute inorganic acid, insoluble in alcohol, practically insoluble in common organic acids
[Mn++].[O-]C([O-])=O
114.95
114.95
3% max. (105°C)
3.12
90%
Amorphous Powder
88% min. (Complexometry)
MnCO3·xH2O
manganese ii carbonate hydrate, co3.mn.h2o, manganese 2+ hydrate carbonate, manganese 2+ ion hydrate carbonate
XMWCXZJXESXBBY-UHFFFAOYSA-L
manganese(2+) carbonate
24848673
90%
Plastic bottle
3.1200g/mL

Chemical Identifiers

34156-69-9
114.95
XMWCXZJXESXBBY-UHFFFAOYSA-L
24848673
[Mn++].[O-]C([O-])=O
CMnO3
MFCD00149794
manganese ii carbonate hydrate, co3.mn.h2o, manganese 2+ hydrate carbonate, manganese 2+ ion hydrate carbonate
manganese(2+) carbonate
Safety and Handling

Safety and Handling

HYGROSCOPIC

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only