missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese(II) carbonate, 99.9% (metals basis excluding Na), Alfa Aesar™

Manufacturer:  Alfa Aesar 14344A1

 View more versions of this product

Catalog No. AA14344A1



Specifications

Specifications

Manganese(II) carbonate
99.9% (metals basis excluding Na)
MFCD00011116
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
C(=O)([O-])[O-].[Mn+2]
114.946
114.95
Odorless
1kg
598-62-9
CMnO3
Light brown to violet powder
XMWCXZJXESXBBY-UHFFFAOYSA-L
manganese(2+);carbonate
11726
Powder
Brown to Violet

Chemical Identifiers

598-62-9
114.946
XMWCXZJXESXBBY-UHFFFAOYSA-L
11726
C(=O)([O-])[O-].[Mn+2]
CMnO3
MFCD00011116
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
manganese(2+);carbonate
Safety and Handling

Safety and Handling

EINECSNumber : 209-942-9

Documents