missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganese(II) bromide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04444318
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Manganese (II) bromide hydrate | |
10031-20-6 | |
Crystalline | |
(metals basis) | |
MFCD00149791 | |
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide | |
O.O.O.O.[Mn++].[Br-].[Br-] | |
286.81 | |
99.9% | |
Hygroscopic |
64.3°C (decomposition) | |
Pink | |
50 g | |
Br2H8MnO4 | |
14,5725 | |
HHDPJRSXPOGIOP-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dibromide | |
214.76 (Anhydrous) | |
Odorless |
Chemical Identifiers
10031-20-6 | |
286.81 | |
HHDPJRSXPOGIOP-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dibromide |
Br2H8MnO4 | |
MFCD00149791 | |
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide | |
O.O.O.O.[Mn++].[Br-].[Br-] |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 236-591-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only