missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1080206

 View more versions of this product

Catalog No. AA1080206



Specifications

Specifications

Manganese(II) acetate tetrahydrate
(metals basis)
Mn(OOCCH3)2·4H2O
14,5724
CESXSDZNZGSWSP-UHFFFAOYSA-L
manganese(2+);diacetate;tetrahydrate
93021
Crystalline Aggregates
Puratronic™
Pungent
80°C
6156-78-1
C4H14MnO8
MFCD00062552
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese ii acetate tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mn+2]
245.086
245.09 (173.03 Anhydrous)
99.999%
1.589g/mL
Pink
5g

Chemical Identifiers

6156-78-1
245.086
CESXSDZNZGSWSP-UHFFFAOYSA-L
93021
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mn+2]
C4H14MnO8
MFCD00062552
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese ii acetate tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
manganese(2+);diacetate;tetrahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AI5775000

TSCA : TSCA

Documents