Please login to your online account to display your discounted pricing

Manganese(II) acetate, +98%, anhydrous, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC389141000

 View more versions of this product

Catalog No. AC389141000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Powder
Formula Weight 173.03
Color Grayish-beige to light brown
Quantity 100g
Merck Index 15, 5789
Chemical Name or Material Manganese(II) acetate
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98+%
Molecular Formula C4H6MnO4
Linear Formula Mn(OC(O)CH3)2
MDL Number MFCD00013039
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Suspected of damaging fertility.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 638-38-0

Specifications

Chemical Name or Material Manganese(II) acetate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H6MnO4
Linear Formula Mn(OC(O)CH3)2
Formula Weight 173.03
MDL Number MFCD00013039
Accessories

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Suspected of damaging fertility.

SDS & Safety Documents