Please login to your online account to display your discounted pricing

Manganese(II) acetate, +98%, anhydrous, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC389140250

 View more versions of this product

Catalog No. AC389140250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Color Grayish-beige to light brown
Formula Weight 173.03
Physical Form Powder
Quantity 25g
Merck Index 15, 5789
CAS 638-38-0
Linear Formula Mn(OC(O)CH3)2
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Suspected of damaging fertility.
Assay Percent Range 98+%
MDL Number MFCD00013039
Packaging Glass bottle
Chemical Name or Material Manganese(II) acetate
Infrared Spectrum Authentic
Molecular Formula C4H6MnO4

Specifications

Chemical Name or Material Manganese(II) acetate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H6MnO4
Linear Formula Mn(OC(O)CH3)2
Formula Weight 173.03
MDL Number MFCD00013039
Accessories

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Suspected of damaging fertility.

SDS & Safety Documents