Learn More
Manganese(II) acetate, 98+%, anhydrous, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 389140050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Manganese(II) acetate | |
5g | |
C4H6MnO4 | |
MFCD00013039 | |
manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y | |
UOGMEBQRZBEZQT-UHFFFAOYSA-L | |
manganese(2+);diacetate | |
12525 | |
98+% | |
Glass bottle |
638-38-0 | |
98% min. (Complexometry) | |
Mn(OC(O)CH3)2 | |
15, 5789 | |
Solubility in water: very soluble | |
[Mn++].CC([O-])=O.CC([O-])=O | |
173.03 | |
173.03 | |
Authentic |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
EINECSNumber : 211-334-3
RUO â Research Use Only