Learn More
Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 038607A7
Description
Used as paint driers. Accelerating curing agent for unsaturated polyester. Also used as a intermediate for dyestuff, pharmaceutical, syntheses material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Manganese(II) 2-ethylhexanoate | |
Brown | |
40°C (104°F) | |
C16H30MnO4 | |
5 kg | |
manganese bis 2-ethylhexanoate, manganese 2-ethylhexanoate, manganese 2+ bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, manganese 2+ salt, hexanoic acid, 2-ethyl-, manganese 2+ salt 2:1, hexanoic acid, 2-ethyl-, manganese salt, hexanoic acid, 2-ethyl-, manganese salt 1:?, manganese tallate 2-ethylhexanoate, manganous 2-ethylhexanoate, mmanganese 2-ethylhexanoate | |
FHRAKXJVEOBCBQ-UHFFFAOYSA-L | |
2-ethylhexanoate;manganese(2+) | |
114521 | |
40% w/w in min.eral spirits, 6% Mn |
13434-24-7 | |
0.897 g/mL | |
Petroleum-like | |
MFCD00058698 | |
UN1268 | |
Soluble in organic solvents. Not miscible or difficult to mix with water. | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Mn+2] | |
341.35 | |
341.35 | |
Liquid |
Chemical Identifiers
13434-24-7 | |
341.35 | |
FHRAKXJVEOBCBQ-UHFFFAOYSA-L | |
114521 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Mn+2] |
C16H30MnO4 | |
MFCD00058698 | |
manganese bis 2-ethylhexanoate, manganese 2-ethylhexanoate, manganese 2+ bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, manganese 2+ salt, hexanoic acid, 2-ethyl-, manganese 2+ salt 2:1, hexanoic acid, 2-ethyl-, manganese salt, hexanoic acid, 2-ethyl-, manganese salt 1:?, manganese tallate 2-ethylhexanoate, manganous 2-ethylhexanoate, mmanganese 2-ethylhexanoate | |
2-ethylhexanoate;manganese(2+) |
Safety and Handling
GHS H Statement
H304-H361-H226
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P281-P301+P310-P303+P361+P353-P308+P313-P331-P370+P378q-P501c
H226-H304-H340-H350-H372
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only