missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese Chloride, 1.0M Solution (Clear, Lt.-Pink Solution/Molecular Biology), Fisher BioReagents

$173.03 - $173.03

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
Synonym acmc-20akkt, ksc171o6h, manganese 2+ ion dichloride, manganese chloride, manganese ii chloride, manganese ii chloride anhydrous crystalline, manganesechloride, manganous chloride, anhydr, manganous chloride,anhydrous
PubChem CID 10313134
ChEBI CHEBI:63041
IUPAC Name manganese(2+);dichloride
SMILES [Cl-].[Cl-].[Mn+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP541-100
SDS View Documents
Fisher BioReagents
FLBP541100
100mL Poly Bottle Each for $173.03
Case of 6 Each for $1,038.18
 
Description

Description

Manganese Chloride is used for dC and dG tailing of DNA.
Specifications

Chemical Identifiers

7773-01-5
197.90
acmc-20akkt, ksc171o6h, manganese 2+ ion dichloride, manganese chloride, manganese ii chloride, manganese ii chloride anhydrous crystalline, manganesechloride, manganous chloride, anhydr, manganous chloride,anhydrous
CHEBI:63041
[Cl-].[Cl-].[Mn+2]
Cl2H8MnO4
CNFDGXZLMLFIJV-UHFFFAOYSA-L
10313134
manganese(2+);dichloride

Specifications

7773-01-5,7732-18-5
0.99 to 1.01M
acmc-20akkt, ksc171o6h, manganese 2+ ion dichloride, manganese chloride, manganese ii chloride, manganese ii chloride anhydrous crystalline, manganesechloride, manganous chloride, anhydr, manganous chloride,anhydrous
[Cl-].[Cl-].[Mn+2]
manganese(2+);dichloride
10313134
Liquid
5ppm max.
Poly Bottle
Red
100mL
Cl2H8MnO4
15, 5793
CNFDGXZLMLFIJV-UHFFFAOYSA-L
DNase free
197.90
CHEBI:63041
Molecular Biology
Pass Test
Protease free
4
Manganese Chloride, 1.0M Solution
Safety and Handling

Safety and Handling

Recommended Storage : RT