Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese bis(trifluoromethylsulfonyl)imide

Chemical Identifiers

Chemical Name or Material Manganese bis(trifluoromethylsulfonyl)imide
CAS 207861-55-0
Linear Formula [(CF3SO2)2N]2Mn
Formula Weight 615.21
MDL Number MFCD23380173
UN Number UN3261
Synonym Manganese bis(trifluoromethanesulfonyl)imide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4717903

  SDS
alfa aesar™
4717903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4717906

  SDS
alfa aesar™
4717906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47179MD

  SDS
alfa aesar™
47179MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 615.21
Chemical Name or Material Manganese bis(trifluoromethylsulfonyl)imide
Physical Form Solid
Sensitivity Hygroscopic
Linear Formula [(CF3SO2)2N]2Mn
MDL Number MFCD23380173
CAS 207861-55-0
UN Number UN3261
Synonym Manganese bis(trifluoromethanesulfonyl)imide
Color Off-white
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.