Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese antimonide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Manganese antimonide
CAS 12032-97-2
Linear Formula Mn2Sb
Formula Weight 231.63
MDL Number MFCD00151354
UN Number UN1549
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3639006

  SDS
alfa aesar™
3639006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3639014

  SDS
alfa aesar™
3639014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 231.63
Chemical Name or Material Manganese antimonide
TSCA TSCA
Particle Size 10mm & down
Assay 99.5%
Assay Percent Notes (metals basis)
Linear Formula Mn2Sb
MDL Number MFCD00151354
CAS 12032-97-2
UN Number UN1549
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.