Please login to your online account to display your discounted pricing

Alfa Aesar™ Manganese aluminide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Manganese aluminide
CAS 12253-13-3
Formula Weight 135.88
MDL Number MFCD00213943
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3638406

  SDS
alfa aesar™
3638406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3638414

  SDS
alfa aesar™
3638414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 135.88
Quantity 5g
Assay 99.5%
TSCA TSCA
Assay Percent Notes (metals basis)
CAS 12253-13-3
MDL Number MFCD00213943
Chemical Name or Material Manganese aluminide
Particle Size 10mm & down