missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Manganese, AAS standard solution, Specpure™, Mn 1000μg/mL, Alfa Aesar™

Manufacturer:  Alfa Aesar 88078AE

 View more versions of this product

Catalog No. AA88078AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Manganese
Mn
Nitric acid: 5%; Manganese: 0.1%; Water: 94.9%
[Mn]
54.938
CHEBI:35154
17mm Hg at 20°C
100mL
7439-96-5,7697-37-2,7732-18-5
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
23930
Liquid
Colorless

Chemical Identifiers

7439-96-5
54.938
PWHULOQIROXLJO-UHFFFAOYSA-N
CHEBI:35154
[Mn]
Mn
MFCD00011111
23930
manganese
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

SDS Documents