missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganese AA Standard, 1000 ppm Mn in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company AMN1KN500
Specifications
Manganese AA Standard | |
7439-96-5,7647-01-0,7732-18-5 | |
500 mL | |
98.0,4.04,0.1 | |
1,000ppm Mn | |
MFCD00011111 | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
Liquid |
In 3% Nitric Acid | |
Natural Poly Bottle | |
94.0,3.88,0.09 | |
Colorless | |
Mn | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
1 mL = 1 mg Mn |
Chemical Identifiers
7439-96-5 | |
54.94 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
CHEBI:35154 | |
[Mn] |
Mn | |
MFCD00011111 | |
23930 | |
manganese |