Please login to your online account to display your discounted pricing

Manganese, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$139.42 - $352.29

Chemical Identifiers

Chemical Name or Material Manganese, 1000μg/mL (0.10% w/v)
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Mn
Formula Weight 54.94
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003281

 
avantor performance
579304
Poly Bottle 100mL Each for $139.42

02003282

 
avantor performance
645804
Narrow Mouth Poly Bottle 150mL Each for $352.29
Description & Specifications

Specifications

Formula Weight 54.94
Quantity 100mL
Packaging Poly Bottle
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Specific Gravity 1L = 1.01kg
Molecular Formula Mn
Chemical Name or Material Manganese, 1000μg/mL (0.10% w/v)