Please login to your online account to display your discounted pricing

Mandelonitrile (Tech.), ACROS Organics™

Chemical Identifiers

Chemical Name or Material Mandelonitrile
Name Note tech.
CAS 532-28-5
Purity Grade Technical Grade
Assay ≥80% (typically)
Molecular Formula C8H7NO
Linear Formula C6H5CH(OH)CN
Formula Weight 133.15
MDL Number MFCD00004487
Synonym α-Hydroxyphenylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC152090250

  SDS
thermo fisher scientific
AC152090250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC152095000

  SDS
thermo fisher scientific
152095000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥80% (typically)
Formula Weight 133.15
Purity Grade Technical Grade
Physical Form Liquid
Quantity 25mL
Color Yellow to brown
EINECS Number 208-532-7
RTECS Number OO8400000
TSCA TSCA
Density 1.11g/mL
Beilstein 10, 193
Merck Index 15, 5783
Refractive Index 1.5305 to 1.5335 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00004487
Synonym α-Hydroxyphenylacetonitrile
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Linear Formula C6H5CH(OH)CN
Molecular Formula C8H7NO
Chemical Name or Material Mandelonitrile
CAS 532-28-5
Name Note tech.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Toxic in contact with skin.
Toxic if swallowed.