Please login to your online account to display your discounted pricing

Mandelonitrile (Tech.), ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  152095000

 View more versions of this product

Catalog No. AC152095000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥80% (typically)
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 133.15
Quantity 500mL
Color Yellow to brown
Beilstein 10, 193
Merck Index 15, 5783
EINECS Number 208-532-7
TSCA TSCA
Density 1.11g/mL
RTECS Number OO8400000
Chemical Name or Material Mandelonitrile
Name Note tech.
Molecular Formula C8H7NO
Infrared Spectrum Authentic
Linear Formula C6H5CH(OH)CN
MDL Number MFCD00004487
CAS 532-28-5
Synonym α-Hydroxyphenylacetonitrile
Refractive Index 1.5305 to 1.5335 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Mandelonitrile
Name Note tech.
CAS 532-28-5
Purity Grade Technical Grade
Assay ≥80% (typically)
Molecular Formula C8H7NO
Linear Formula C6H5CH(OH)CN
Formula Weight 133.15
MDL Number MFCD00004487
Synonym α-Hydroxyphenylacetonitrile
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Toxic in contact with skin.
Toxic if swallowed.

SDS & Safety Documents