Please login to your online account to display your discounted pricing

Alfa Aesar™ Mandelonitrile, tech. 80%

Chemical Identifiers

Chemical Name or Material Mandelonitrile
CAS 532-28-5
Purity Grade Technical Grade
Molecular Formula C8H7NO
Formula Weight 133.15
MDL Number MFCD00004487
UN Number UN3276
Beilstein 2207122
Merck Index 14,5719
Synonym Benzaldehyde cyanohydrin; alpha-Hydroxyphenylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0869814

  SDS
alfa aesar™
L0869814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0869822

  SDS
alfa aesar™
L0869822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0869836

  SDS
alfa aesar™
L0869836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.15
Chemical Name or Material Mandelonitrile
MDL Number MFCD00004487
Health Hazard 1 Danger
Assay 80%
Health Hazard 2 GHS H Statement
H301-H318-H315
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
Synonym Benzaldehyde cyanohydrin; alpha-Hydroxyphenylacetonitrile
Beilstein 2207122
UN Number UN3276
Melting Point −10°C
CAS 532-28-5
Molecular Formula C8H7NO
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number OO8400000
TSCA TSCA
Boiling Point 168° to 170°C
Purity Grade Technical Grade
Merck Index 14,5719
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Quantity 25g
Flash Point 97°C (206°F)
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.