Learn More
Mandelic acid hydrazide, 97%, Thermo Scientific Chemicals
$102.26 - $345.96
Chemical Identifiers
CAS | 2443-66-5 |
---|---|
Molecular Formula | C8H10N2O2 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00038133 |
InChI Key | FWTGUGVETHVGTL-UHFFFAOYSA-N |
Synonym | mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667 |
PubChem CID | 73126 |
IUPAC Name | 2-hydroxy-2-phenylacetohydrazide |
SMILES | C1=CC=C(C=C1)C(C(=O)NN)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1165306
|
Thermo Scientific Chemicals
L1165306 |
5 g |
Each for $102.26
|
|
|||||
AAL1165314
|
Thermo Scientific Chemicals
L1165314 |
25 g |
Each for $345.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2443-66-5 | |
166.18 | |
FWTGUGVETHVGTL-UHFFFAOYSA-N | |
73126 | |
C1=CC=C(C=C1)C(C(=O)NN)O |
C8H10N2O2 | |
MFCD00038133 | |
mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667 | |
2-hydroxy-2-phenylacetohydrazide |
Specifications
2443-66-5 | |
C8H10N2O2 | |
5 g | |
mandelic acid hydrazide, mandelhydrazide, mandelhydrazine, mandelic acid, hydrazide, mandelic hydrazide, hydroxy-phenyl-acetic acid hydrazide, 2-???-1-phenylethan-1-ol, mandelohydrazide, asischem d13400, akos bc-1667 | |
C1=CC=C(C=C1)C(C(=O)NN)O | |
166.18 | |
166.18 | |
Mandelic acid hydrazide |
128°C to 131°C | |
MFCD00038133 | |
2096197 | |
FWTGUGVETHVGTL-UHFFFAOYSA-N | |
2-hydroxy-2-phenylacetohydrazide | |
73126 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
RTECSNumber : OO7450000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only