Please login to your online account to display your discounted pricing

Alfa Aesar™ Maltulose monohydrate, 96%

$251.66 - $251.66

Chemical Identifiers

Chemical Name or Material Maltulose monohydrate
CAS 207511-09-9
Molecular Formula C12H22O11·H2O
Formula Weight 360.32g/mol (342.30g/mol Anhydrous)
MDL Number MFCD00217057
Synonym 4-O-alpha-D-Glucopyranosyl-D-fructose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2114106

 
alfa aesar™
B21141-06
5g Each for $251.66

AAB2114103

 
alfa aesar™
B21141-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point ≈125°C (dec.)
Quantity 5g
Chemical Name or Material Maltulose monohydrate
CAS 207511-09-9
Assay 96%
Molecular Formula C12H22O11·H2O
Formula Weight 360.32g/mol (342.30g/mol Anhydrous)
MDL Number MFCD00217057
Synonym 4-O-alpha-D-Glucopyranosyl-D-fructose