Please login to your online account to display your discounted pricing

Maltose OR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Maltose
Name Note Monohydrate
CAS 6363-53-7
Molecular Formula C12H22O11·H2O
Formula Weight 360.32g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK1881250

 
avantor performance
188158
Glass 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Glass
Quantity 250g
Chemical Name or Material Maltose
Name Note Monohydrate
CAS 6363-53-7
Molecular Formula C12H22O11·H2O
Formula Weight 360.32g/mol