Please login to your online account to display your discounted pricing

Thermo Scientific™ Maltose Binding Protein Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ Maltose Binding Protein Ab-2, Mouse Monoclonal Antibody.

$240.37 - $1,022.29

Specifications

Antigen Maltose Binding Protein Ab-2
Clone R29
Applications Immunofluorescence
Applications Western Blotting
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS765P

 
thermo scientific™
MS765P
200μg/mL 1mL Each for $947.06

MS765P0

 
thermo scientific™
MS765P0
200μg/mL 100μL Each for $240.37

MS765P1

 
thermo scientific™
MS765P1
200μg/mL 500μL Each for $667.88

MS765P1ABX

 
thermo scientific™
MS765P1ABX
1mg/mL 100μL Each for $764.00

MS765PABX

 
thermo scientific™
MS765PABX
1mg/mL 200μL Each for $1,022.29
Description & Specifications

Specifications

Antigen Maltose Binding Protein Ab-2
Clone R29
Applications Immunofluorescence
Applications Western Blotting
Applications Immunofluorescence
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity All
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Epitope Tags
Type Primary
Immunogen Purified recombinant MBP fusion protein
Isotype IgG1

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ Maltose Binding Protein Ab-2, Mouse Monoclonal Antibody.

The approach of epitope tagging offers an easy and universal strategy for the identification and purification of proteins derived by recombinant DNA technology. The insertion of a MBP tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the MBP-tagged product. Cleavage by factor Xa separates MBP from its partner protein.

Host Species: Mouse

Clone: R29

Isotype: IgG1

Species Reactivity: Bacteria. Proteins from all species which are tagged with MBP.

Epitope: Not determined

Immunogen: Purified recombinant MBP fusion protein

Molecular Weight: 53kDa

Positive Control: MBP fusion protein in bacteria, bacterial lysate, or cells and tissues transfected with an MBP fusion protein expressing vectors

Cellular Localization: Depends upon the localization of the parent protein tagged with MBP

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO