Please login to your online account to display your discounted pricing

Thermo Scientific™ Maltose Binding Protein Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in experiments with the Thermo Scientific™ Maltose Binding Protein Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™  MS765P1ABX

 View more versions of this product

Catalog No. MS765P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity All
Host Species Mouse
Immunogen Purified recombinant MBP fusion protein
Format Concentrated
Regulatory Status RUO
Isotype IgG1
Applications Immunofluorescence
Applications Western Blotting
Quantity 100µL
Antigen Maltose Binding Protein Ab-2
Clone R29
Concentration 1mg/mL
Research Discipline Epitope Tags
Type Primary
Conjugate Unlabeled
Classification Monoclonal

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ Maltose Binding Protein Ab-2, Mouse Monoclonal Antibody.

The approach of epitope tagging offers an easy and universal strategy for the identification and purification of proteins derived by recombinant DNA technology. The insertion of a MBP tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the MBP-tagged product. Cleavage by factor Xa separates MBP from its partner protein.

Host Species: Mouse

Clone: R29

Isotype: IgG1

Species Reactivity: Bacteria. Proteins from all species which are tagged with MBP.

Epitope: Not determined

Immunogen: Purified recombinant MBP fusion protein

Molecular Weight: 53kDa

Positive Control: MBP fusion protein in bacteria, bacterial lysate, or cells and tissues transfected with an MBP fusion protein expressing vectors

Cellular Localization: Depends upon the localization of the parent protein tagged with MBP

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO