Learn More
Malonyl dichloride, 94%, Thermo Scientific Chemicals
$231.71 - $732.90
Chemical Identifiers
CAS | 1663-67-8 |
---|---|
Molecular Formula | C3H2Cl2O2 |
Molecular Weight (g/mol) | 140.947 |
MDL Number | MFCD00000735 |
InChI Key | SXYFKXOFMCIXQW-UHFFFAOYSA-N |
Synonym | malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride |
PubChem CID | 74269 |
IUPAC Name | propanedioyl dichloride |
SMILES | C(C(=O)Cl)C(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5986814
|
Thermo Scientific Chemicals
H5986814 |
25 g |
Each for $231.71
|
|
|||||
AAH5986822
|
Thermo Scientific Chemicals
H5986822 |
100 g |
Each for $732.90
|
|
|||||
Description
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMalonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Decomposes in water.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
Chemical Identifiers
1663-67-8 | |
140.947 | |
SXYFKXOFMCIXQW-UHFFFAOYSA-N | |
74269 | |
C(C(=O)Cl)C(=O)Cl |
C3H2Cl2O2 | |
MFCD00000735 | |
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride | |
propanedioyl dichloride |
Specifications
1663-67-8 | |
53°C to 55°C (19 mmHg) | |
C3H2Cl2O2 | |
CH2(COCl)2 | |
25 g | |
774044 | |
Decomposes in water. | |
C(C(=O)Cl)C(=O)Cl | |
140.947 | |
140.95 | |
Malonyl dichloride |
1.449 | |
47°C (117°F) | |
1.465 | |
MFCD00000735 | |
UN2920 | |
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride | |
SXYFKXOFMCIXQW-UHFFFAOYSA-N | |
propanedioyl dichloride | |
74269 | |
97% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 216-772-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only