missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonyl Chloride, TCI America™

Manufacturer:  TCI America M088225G

 View more versions of this product

Catalog No. M088225G



Specifications

Specifications

Malonyl Chloride
Undesignated
1663-67-8
MFCD00000735
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
C(C(=O)Cl)C(=O)Cl
140.947
140.95
≥97.0% (T)
60°C
25g
C3H2Cl2O2
2920
SXYFKXOFMCIXQW-UHFFFAOYSA-N
propanedioyl dichloride
74269
Clear Liquid at 20°C

Chemical Identifiers

1663-67-8
140.947
SXYFKXOFMCIXQW-UHFFFAOYSA-N
74269
C(C(=O)Cl)C(=O)Cl
C3H2Cl2O2
MFCD00000735
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
propanedioyl dichloride
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents