missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonyl Chloride, TCI America™

$93.20 - $161.95

Chemical Identifiers

CAS 1663-67-8
Molecular Formula C3H2Cl2O2
Molecular Weight (g/mol) 140.947
MDL Number MFCD00000735
InChI Key SXYFKXOFMCIXQW-UHFFFAOYSA-N
Synonym malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
PubChem CID 74269
IUPAC Name propanedioyl dichloride
SMILES C(C(=O)Cl)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M088210G View Documents TCI America
M088210G
10g Each for $93.20
 
M088225G View Documents TCI America
M088225G
25g Each for $161.95
 
Specifications

Chemical Identifiers

1663-67-8
140.947
SXYFKXOFMCIXQW-UHFFFAOYSA-N
74269
C(C(=O)Cl)C(=O)Cl
C3H2Cl2O2
MFCD00000735
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
propanedioyl dichloride

Specifications

60°C
10g
C3H2Cl2O2
2920
SXYFKXOFMCIXQW-UHFFFAOYSA-N
propanedioyl dichloride
74269
Clear Liquid at 20°C
Malonyl Chloride
Undesignated
1663-67-8
MFCD00000735
malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
C(C(=O)Cl)C(=O)Cl
140.947
140.95
≥97.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator