missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malononitrile 98.0+%, TCI America™
Supplier: TCI America M0033100G
Specifications
| Malononitrile | |
| 220°C | |
| MFCD00001883 | |
| 2647 | |
| CUONGYYJJVDODC-UHFFFAOYSA-N | |
| propanedinitrile | |
| 8010 | |
| 66.06 | |
| Crystalline Lumps |
| 109-77-3 | |
| C3H2N2 | |
| 100 g | |
| malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano | |
| C(C#N)C#N | |
| 66.063 | |
| CHEBI:33186 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 109-77-3 | |
| 66.063 | |
| CUONGYYJJVDODC-UHFFFAOYSA-N | |
| 8010 | |
| propanedinitrile |
| C3H2N2 | |
| MFCD00001883 | |
| malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano | |
| CHEBI:33186 | |
| C(C#N)C#N |
Safety and Handling
EINECSNumber : (2)-1511
RTECSNumber : OO3150000
TSCA : Yes
Recommended Storage : Refrigerator