missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Malononitrile, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 125271000
Specifications
Malononitrile | |
112°C | |
Glass bottle | |
220.0°C | |
30.0°C to 34.0°C | |
99% | |
98.5% min. (GC) | |
NCCH2CN | |
02,589 | |
15,5775 | |
Solubility in water: 133g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone | |
C(C#N)C#N | |
66.06 | |
CHEBI:33186 | |
Crystalline Low Melting Mass |
1.0400g/mL | |
Authentic | |
1.04 | |
Brown-Yellow to White | |
100g | |
109-77-3 | |
C3H2N2 | |
MFCD00001883 | |
04,317 | |
cyanoacetonitrile, dicyanmethane, dicyanomethane, malonic acid dinitrile, malonic dinitrile, malonodinitrile, malononitrile, methane, dicyano, methylene cyanide, methylenedinitrile | |
CUONGYYJJVDODC-UHFFFAOYSA-N | |
propanedinitrile | |
8010 | |
66.06 | |
99% |
Chemical Identifiers
109-77-3 | |
66.06 | |
CUONGYYJJVDODC-UHFFFAOYSA-N | |
8010 | |
propanedinitrile |
C3H2N2 | |
MFCD00001883 | |
cyanoacetonitrile, dicyanmethane, dicyanomethane, malonic acid dinitrile, malonic dinitrile, malonodinitrile, malononitrile, methane, dicyano, methylene cyanide, methylenedinitrile | |
CHEBI:33186 | |
C(C#N)C#N |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
GHS Signal Word: Danger
EINECSNumber : 203-703-2
RTECSNumber : OO3150000
TSCA : TSCA