Please login to your online account to display your discounted pricing

Malonoben, Tocris


Oxidative phosphorylation uncoupler

Quantity 50mg

Manufacturer: r & d systems  RDS047950

 View more versions of this product

Catalog No. 047950

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 282.38
Chemical Name or Material Malonoben
Assay Percent Range >98%
Molecular Formula C18H22N2O
CAS 10537-47-0
Synonym 3, 5-Di-tert-butyl-4-hydroxybenzylidenemalononitrile
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Malonoben
Assay Percent Range >98%
Molecular Formula C18H22N2O
Formula Weight 282.38
Synonym 3, 5-Di-tert-butyl-4-hydroxybenzylidenemalononitrile
Accessories

Store at Room Temperature