missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonic acid, Reagent Grade, 99.5+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03171536

 View more versions of this product

Catalog No. AA3171536



Description

Description

Malonic acid is used to prepare barbiturates. Its derivative diethylmalonate is used in Knoevenagel condensation reactions as well as reacts with acetone to form Meldrum's acid. It is used to control acidity, preservative additive for foods and as an excipient in pharmaceutical formulation. It acts as a precursor to polyesters and as an active component in alkyd resins. Further, it is used as a building block in chemical synthesis. In addition to this, it is used to prepare starch-based resin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Malonic acid
132°C to 136°C (decomposition)
1.62 g/mL
C3H4O4
MFCD00002707
1751370
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
OFOBLEOULBTSOW-UHFFFAOYSA-N
propanedioic acid
867
104.06
Reagent
141-82-2
White
157°C (314°F)
H2C(CO2H)2
500 g
14,5710
Soluble in water,sodium hydroxide and alcohol; Slightly soluble in pyridine and ether.
C(C(=O)O)C(=O)O
104.061
CHEBI:30794
≥99.5%
Crystalline

Chemical Identifiers

141-82-2
104.061
OFOBLEOULBTSOW-UHFFFAOYSA-N
867
propanedioic acid
C3H4O4
MFCD00002707
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
CHEBI:30794
C(C(=O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H318-H335

EINECSNumber : 205-503-0

RTECSNumber : OO8175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only