missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Malonic Acid Reagent, For Ozone, LabChem™

$53.63 - $53.63

Chemical Identifiers

CAS 7732-18-5,141-82-2
CAS Max % 95,5
Linear Formula C3H4O4
Formula Weight 104.06
Chemical Name or Material Malonic Acid Reagent, For Ozone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC165207 Lab Chem
LC165207
125mL Each for $53.63
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,141-82-2
C3H4O4
Malonic Acid Reagent, For Ozone
95,5
104.06

Specifications

7732-18-5,141-82-2
Liquid
104.06
Passes Test
Soluble in water
Room Temperature
Clear, colorless
Malonic Acid Reagent, For Ozone
95,5
C3H4O4
1.05g/mL
Poly Bottle
1.05g/mL
Carbon monoxide; Carbon dioxide
125mL

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.

Danger