Please login to your online account to display your discounted pricing

Malonic acid disodium salt monohydrate, 99%, ACROS Organics™

$58.68 - $505.67

Chemical Identifiers

Chemical Name or Material Malonic acid disodium salt monohydrate
CAS 26522-85-0
CAS 26522-85-0
Assay ≥98.5% (on dry substance) (Total base)
Assay Percent Range 99%
Molecular Formula C3H2Na2O4·xH2O
Linear Formula CH2(COONa)2·H2O
Formula Weight 166.05
MDL Number MFCD00149114
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC413505000

  SDS
thermo fisher scientific
AC413505000
Plastic bottle 500g Each for $505.67

AC413500250

  SDS
thermo fisher scientific
413500250
Plastic bottle 25g Each for $58.68

AC413501000

  SDS
fisher scientific acros
413501000
Plastic bottle 100g Each for $110.30
Description & Specifications

Specifications

Assay ≥98.5% (on dry substance) (Total base)
Physical Form Wax
Formula Weight 166.05
Quantity 500g
Color Yellow
CAS 26522-85-0
Infrared Spectrum Authentic
Water 7 to 13% (K.F.)
Molecular Formula C3H2Na2O4·xH2O
Assay Percent Range 99%
MDL Number MFCD00149114
Chemical Name or Material Malonic acid disodium salt monohydrate
Packaging Plastic bottle
Linear Formula CH2(COONa)2·H2O
CAS 26522-85-0